Assay Details
Target Gene Details
Entrez Gene ID: | 60491 |
Gene Name: | NGG1 interacting factor 3 like 1 |
Gene Aliases: |
ALS2CR1, CALS-7, MDS015 |
Location: |
Chr.2:200889327-200903932 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NIF3L1 | NM_001136039.2 | NP_001129511.1 | ||
NM_001142355.1 | NP_001135827.1 | |||
NM_001142356.1 | NP_001135828.1 | |||
NM_021824.3 | NP_068596.2 | |||
XM_005246742.3 | XP_005246799.1 | |||
XM_011511582.1 | XP_011509884.1 | |||
XM_011511583.1 | XP_011509885.1 | |||
XM_011511584.1 | XP_011509886.1 | |||
XM_011511585.1 | XP_011509887.1 | |||
XM_017004630.1 | XP_016860119.1 | |||
AB038949.1 | BAB32499.1 | |||
AF060513.1 | AAG43131.1 | |||
AF182416.1 | AAG14952.1 | |||
AF283538.1 | AAG44846.1 | |||
AK023378.1 | BAB14551.1 | |||
AK225905.1 | ||||
AK314709.1 | ||||
AL834430.1 | CAH56370.1 | |||
AY251943.1 | AAP84063.1 | |||
AY251944.1 | AAP84064.1 | |||
BC007654.2 | AAH07654.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3109 | Chr.2:200896870 - 200942486 on Build GRCh38 | Deletion | ORC2 NIF3L1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |