Assay Details
Target Gene Details
Entrez Gene ID: | 8440 |
Gene Name: | NCK adaptor protein 2 |
Gene Aliases: |
GRB4, NCKbeta |
Location: |
Chr.2:105744649-105894274 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NCK2 | NM_001004722.3 | NP_001004722.1 | ||
NM_003581.4 | NP_003572.2 | |||
XM_006712797.3 | XP_006712860.1 | |||
XM_011511991.2 | XP_011510293.1 | |||
XM_017005103.1 | XP_016860592.1 | |||
XM_017005105.1 | XP_016860594.1 | |||
BG480643.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834321 | Chr.2:105663236 - 105841053 on Build GRCh38 | Loss | NCK2 |
dgv1159e212 | Chr.2:105767281 - 105773234 on Build GRCh38 | Loss | NCK2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |