Assay Details
Target Gene Details
Entrez Gene ID: | 102724691 |
Gene Name: | uncharacterized LOC102724691 |
Gene Aliases: |
- |
Location: |
Chr.2:104936241-105038496 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC102724691 | NR_110603.1 | |||
BX109606.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)