Assay Details
Target Gene Details
Entrez Gene ID: | 56655 |
Gene Name: | DNA polymerase epsilon 4, accessory subunit |
Gene Aliases: |
YHHQ1, p12 |
Location: |
Chr.2:74958648-74969732 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POLE4 | NM_019896.2 | NP_063949.2 | ||
AY034104.1 | ||||
BC031331.1 | AAH31331.1 | |||
CA454961.1 | ||||
CF529348.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834262 | Chr.2:74907892 - 75062072 on Build GRCh38 | Gain | LINC01291 TACR1 LOC105374809 LINC01293 POLE4 |
esv2751900 | Chr.2:74722905 - 75041937 on Build GRCh38 | Gain | LINC01291 LOC105374809 LINC01293 POLE4 HK2 |
esv3575258 | Chr.2:74925505 - 75012498 on Build GRCh38 | Gain | LINC01291 LOC105374809 LINC01293 POLE4 |
dgv3872n100 | Chr.2:74917558 - 75013442 on Build GRCh38 | Gain | LINC01291 LOC105374809 LINC01293 POLE4 |
nsv526995 | Chr.2:74916957 - 75012498 on Build GRCh38 | Gain | LINC01291 LOC105374809 LINC01293 POLE4 |
nsv582217 | Chr.2:74736541 - 74993869 on Build GRCh38 | Gain | LINC01291 LOC105374809 LINC01293 POLE4 HK2 |
nsv1001495 | Chr.2:74598036 - 75093218 on Build GRCh38 | Gain | LINC01291 MIR5000 TACR1 LOC105374809 SEMA4F LINC01293 POLE4 M1AP HK2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |