Assay Details
Target Gene Details
Entrez Gene ID: | 53938 |
Gene Name: | peptidylprolyl isomerase like 3 |
Gene Aliases: |
CYPJ |
Location: |
Chr.2:200870907-200889333 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPIL3 | NM_032472.3 | NP_115861.1 | ||
NM_130906.2 | NP_570981.1 | |||
XM_005246651.3 | XP_005246708.1 | |||
XM_005246652.4 | XP_005246709.1 | |||
XM_011511358.2 | XP_011509660.1 | |||
XM_017004352.1 | XP_016859841.1 | |||
XM_017004353.1 | XP_016859842.1 | |||
XM_017004354.1 | XP_016859843.1 | |||
XM_017004355.1 | XP_016859844.1 | |||
XM_017004356.1 | XP_016859845.1 | |||
XM_017004357.1 | XP_016859846.1 | |||
XM_017004358.1 | XP_016859847.1 | |||
XM_017004359.1 | XP_016859848.1 | |||
XM_017004360.1 | XP_016859849.1 | |||
AF146799.1 | AAO64723.1 | |||
AF251049.1 | AAK34939.1 | |||
AF271652.1 | AAG44766.1 | |||
AF414127.1 | AAP97286.1 | |||
AK297804.1 | ||||
AY257468.1 | AAP13527.1 | |||
BC007693.2 | AAH07693.1 | |||
CB961908.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |