Assay Details
Target Gene Details
Entrez Gene ID: | 57624 |
Gene Name: | neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adaptor 2 |
Gene Aliases: |
KIAA1486 |
Location: |
Chr.2:225399710-225703653 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NYAP2 | NM_020864.1 | NP_065915.1 | ||
XM_005246708.3 | XP_005246765.1 | |||
XM_011511524.1 | XP_011509826.1 | |||
XM_017004564.1 | XP_016860053.1 | |||
AB040919.1 | ||||
AK055226.1 | BAB70880.1 | |||
BC131727.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)