Assay Details
Target Gene Details
Entrez Gene ID: | 60509 |
Gene Name: | ATP/GTP binding protein like 5 |
Gene Aliases: |
CCP5, RP75 |
Location: |
Chr.2:27051423-27070622 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AGBL5 | NM_021831.5 | NP_068603.4 | ||
NR_104246.1 | ||||
XM_005264477.2 | XP_005264534.1 | |||
XM_011533011.2 | XP_011531313.1 | |||
XM_011533012.2 | XP_011531314.1 | |||
XM_011533013.2 | XP_011531315.1 | |||
AL833844.1 | CAD38704.1 | |||
BC007415.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv6744n54 | Chr.2:26973431 - 27151141 on Build GRCh38 | Loss | AGBL5 TRA-AGC8-1 ABHD1 AGBL5-AS1 LOC100129724 PRR30 TCF23 OST4 MAPRE3 KHK CGREF1 PREB TMEM214 EMILIN1 |
nsv817673 | Chr.2:27037601 - 27163931 on Build GRCh38 | Gain | AGBL5 PRR30 TCF23 OST4 KHK CGREF1 PREB TRA-AGC8-1 ABHD1 TMEM214 EMILIN1 AGBL5-AS1 |
nsv833603 | Chr.2:27059106 - 27263631 on Build GRCh38 | Loss | CAD AGBL5 ATRAID ABHD1 SLC5A6 PRR30 TCF23 OST4 KHK CGREF1 PREB SLC30A3 EMILIN1 |
nsv455974 | Chr.2:26989775 - 27128411 on Build GRCh38 | Loss | AGBL5 OST4 MAPRE3 KHK CGREF1 TRA-AGC8-1 ABHD1 TMEM214 EMILIN1 AGBL5-AS1 LOC100129724 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |