Assay Details
Target Gene Details
Entrez Gene ID: | 79699 |
Gene Name: | zyg-11 family member B, cell cycle regulator |
Gene Aliases: |
ZYG11 |
Location: |
Chr.1:52726454-52827342 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZYG11B | NM_024646.2 | NP_078922.1 | ||
XM_017002336.1 | XP_016857825.1 | |||
AB051517.1 | ||||
AK296460.1 | ||||
DQ646393.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1001535 | Chr.1:52620368 - 53346539 on Build GRCh38 | Gain | ECHDC2 CPT2 SLC1A7 C1orf123 MIR5095 LRP8 ZYG11A LOC105378732 MAGOH PODN MIR1273G COA7 ZYG11B MIR1273F FAM159A LOC100507564 SCP2 |
nsv829859 | Chr.1:52723470 - 52925273 on Build GRCh38 | Loss | ECHDC2 ZYG11B ZYG11A |
esv2758934 | Chr.1:52660343 - 52823509 on Build GRCh38 | Loss | COA7 ZYG11B FAM159A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |