Assay Details
Target Gene Details
Entrez Gene ID: | 387338 |
Gene Name: | NOP2/Sun RNA methyltransferase family member 4 |
Gene Aliases: |
SHTAP |
Location: |
Chr.1:46340177-46365152 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NSUN4 | NM_001256127.1 | NP_001243056.1 | ||
NM_001256128.1 | NP_001243057.1 | |||
NM_199044.3 | NP_950245.2 | |||
NR_045789.1 | ||||
NR_045790.1 | ||||
NR_045791.1 | ||||
AK021577.1 | BAB13847.1 | |||
AK057420.1 | ||||
AK097524.1 | ||||
AK128066.1 | BAC87258.1 | |||
AK291741.1 | ||||
AK292353.1 | ||||
AK295003.1 | ||||
BC014441.1 | AAH14441.1 | |||
BX360729.2 | ||||
BX648592.1 | ||||
DA799314.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3577837 | Chr.1:46338361 - 46357126 on Build GRCh38 | Loss | NSUN4 |
nsv159955 | Chr.1:46347871 - 46354655 on Build GRCh38 | Deletion | NSUN4 |
nsv829748 | Chr.1:46258836 - 46410450 on Build GRCh38 | Loss | LRRC41 UQCRH RAD54L FAAH NSUN4 |
esv2761743 | Chr.1:45874186 - 46561071 on Build GRCh38 | Gain | DMBX1 LRRC41 LINC01398 MKNK1-AS1 LURAP1 LOC101929626 MKNK1 FAAH POMGNT1 PIK3R3 NSUN4 UQCRH RAD54L FAAHP1 TSPAN1 KNCN MAST2 |
nsv546162 | Chr.1:46341925 - 46405089 on Build GRCh38 | Loss | FAAH NSUN4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |