Assay Details
Target Gene Details
Entrez Gene ID: | 2946 |
Gene Name: | glutathione S-transferase mu 2 (muscle) |
Gene Aliases: |
GST4, GSTM, GSTM2-2, GTHMUS |
Location: |
Chr.1:109668022-109683997 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GSTM2 | NM_001142368.1 | NP_001135840.1 | ||
AK299482.1 | ||||
AK309796.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv258n100 | Chr.1:109658958 - 109700319 on Build GRCh38 | Gain | GSTM1 GSTM2 GSTM4 |
dgv254n100 | Chr.1:109631914 - 109700319 on Build GRCh38 | Gain+Loss | GSTM1 GSTM2 GSTM4 AMPD2 |
dgv15n111 | Chr.1:109664699 - 109703504 on Build GRCh38 | Deletion | GSTM1 GSTM2 GSTM4 |
dgv256n100 | Chr.1:109638683 - 109700319 on Build GRCh38 | Loss | GSTM1 GSTM2 GSTM4 |
nsv824076 | Chr.1:109671310 - 109702964 on Build GRCh38 | Loss | GSTM1 GSTM2 |
nsv946134 | Chr.1:109668513 - 109716699 on Build GRCh38 | Duplication | GSTM1 GSTM5 GSTM2 |
nsv508471 | Chr.1:109674379 - 109692838 on Build GRCh38 | Deletion | GSTM1 GSTM2 |
esv998761 | Chr.1:109673918 - 109701984 on Build GRCh38 | Gain | GSTM1 GSTM2 |
dgv257n100 | Chr.1:109650144 - 109707602 on Build GRCh38 | Gain | GSTM1 GSTM2 GSTM4 |
nsv10617 | Chr.1:109674658 - 109715089 on Build GRCh38 | Gain | GSTM1 GSTM5 GSTM2 |
nsv482081 | Chr.1:109668022 - 109683996 on Build GRCh38 | Gain | GSTM2 |
dgv77e212 | Chr.1:109635559 - 109709330 on Build GRCh38 | Loss | GSTM1 GSTM2 GSTM4 |
dgv3e203 | Chr.1:109676386 - 109710508 on Build GRCh38 | Gain+Loss | GSTM1 GSTM2 |
dgv259n100 | Chr.1:109663341 - 109700319 on Build GRCh38 | Gain+Loss | GSTM1 GSTM2 GSTM4 |
nsv547487 | Chr.1:109674350 - 109713598 on Build GRCh38 | Gain | GSTM1 GSTM5 GSTM2 |
nsv547486 | Chr.1:109657738 - 109690459 on Build GRCh38 | Loss | GSTM1 GSTM2 GSTM4 |
dgv260n100 | Chr.1:109676374 - 109694313 on Build GRCh38 | Loss | GSTM1 GSTM2 |
esv25172 | Chr.1:109644224 - 109716486 on Build GRCh38 | Gain+Loss | GSTM1 GSTM5 GSTM2 GSTM4 |
nsv2310 | Chr.1:109674297 - 109718036 on Build GRCh38 | Deletion | GSTM1 GSTM5 GSTM2 |
esv1003982 | Chr.1:109672531 - 109717528 on Build GRCh38 | Deletion | GSTM1 GSTM5 GSTM2 |
dgv253n100 | Chr.1:109613590 - 109700319 on Build GRCh38 | Gain | GNAT2 GSTM1 GSTM2 GSTM4 AMPD2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |