Assay Details
Target Gene Details
Entrez Gene ID: | 4803 |
Gene Name: | nerve growth factor |
Gene Aliases: |
Beta-NGF, HSAN5, NGFB |
Location: |
Chr.1:115285915-115338253 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NGF | NM_002506.2 | NP_002497.2 | ||
XM_006710663.3 | XP_006710726.1 | |||
XM_011541518.2 | XP_011539820.1 | |||
BC032517.2 | AAH32517.2 | |||
X52599.1 | CAA36832.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv34370 | Chr.1:115181074 - 115625337 on Build GRCh38 | Gain | NGF |
esv26121 | Chr.1:115020681 - 115560070 on Build GRCh38 | Loss | NGF TSPAN2 TSHB |
nsv818389 | Chr.1:115185327 - 115526418 on Build GRCh38 | Gain | NGF |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |