Assay Details
Target Gene Details
Entrez Gene ID: | 353500 |
Gene Name: | bone morphogenetic protein 8a |
Gene Aliases: |
- |
Location: |
Chr.1:39491626-39529869 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BMP8A | NM_181809.3 | NP_861525.2 | ||
XM_006710616.3 | XP_006710679.1 | |||
XM_011541381.2 | XP_011539683.1 | |||
XM_011541382.2 | XP_011539684.1 | |||
XM_017001198.1 | XP_016856687.1 | |||
AY303954.1 | AAP74559.1 | |||
BC131617.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)