Assay Details
Target Gene Details
Entrez Gene ID: | 11240 |
Gene Name: | peptidyl arginine deiminase 2 |
Gene Aliases: |
PAD-H19, PAD2, PDI2 |
Location: |
Chr.1:17066761-17119453 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PADI2 | NM_007365.2 | NP_031391.2 | ||
XM_017000148.1 | XP_016855637.1 | |||
AB023211.1 | ||||
AB030176.1 | BAA82557.1 | |||
AK127006.1 | ||||
AK295783.1 | ||||
BC009701.1 | AAH09701.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428421 | Chr.1:16437838 - 17157486 on Build GRCh38 | Gain+Loss | RNU1-4 FAM231A RNU1-3 CROCC RNU1-2 SDHB MIR3675 LOC100132147 LOC105376805 ATP13A2 LOC102723383 RNU1-1 SPATA21 FAM231B CROCCP2 MFAP2 NECAP2 PADI2 NBPF1 MST1L MST1P2 CROCCP3 ESPNP LOC105376796 |
nsv545690 | Chr.1:17040376 - 17159158 on Build GRCh38 | Gain | PADI2 SDHB |
nsv1145323 | Chr.1:17076286 - 17100733 on Build GRCh38 | Deletion | PADI2 |
nsv834280 | Chr.1:16888218 - 17123006 on Build GRCh38 | Loss | CROCC MFAP2 RNU1-2 PADI2 SDHB LOC105376805 ATP13A2 |
esv3585359 | Chr.1:17077448 - 17111045 on Build GRCh38 | Gain | PADI2 |
esv2422314 | Chr.1:17080898 - 17216627 on Build GRCh38 | Duplication | PADI2 LOC400743 PADI1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |