Assay Details
Target Gene Details
Entrez Gene ID: | 84966 |
Gene Name: | immunoglobin superfamily member 21 |
Gene Aliases: |
- |
Location: |
Chr.1:18107746-18378483 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IGSF21 | NM_032880.4 | NP_116269.3 | ||
XM_011542319.2 | XP_011540621.1 | |||
AK075316.1 | BAC11542.1 | |||
AK294121.1 | ||||
BC007618.1 | AAH07618.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834313 | Chr.1:17955421 - 18162607 on Build GRCh38 | Loss | IGSF21 LOC101927876 |
dgv177n54 | Chr.1:18093650 - 18117498 on Build GRCh38 | Loss | IGSF21 |
nsv1160182 | Chr.1:17655418 - 18282385 on Build GRCh38 | Duplication | ARHGEF10L ACTL8 IGSF21 LOC101927876 |