Assay Details
Target Gene Details
Entrez Gene ID: | 80835 |
Gene Name: | taste 1 receptor member 1 |
Gene Aliases: |
GM148, GPR70, T1R1, TR1 |
Location: |
Chr.1:6555278-6579757 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TAS1R1 | NM_138697.3 | NP_619642.2 | ||
NM_177540.2 | NP_803884.1 | |||
AK292014.1 | ||||
AK293499.1 | ||||
AK315805.1 | ||||
BC136515.1 | ||||
BC136516.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831647 | Chr.1:6525809 - 6587530 on Build GRCh38 | Loss | TAS1R1 NOL9 ZBTB48 |
nsv7880 | Chr.1:6417968 - 6611914 on Build GRCh38 | Gain+Loss | TAS1R1 PLEKHG5 ESPN NOL9 TNFRSF25 KLHL21 ZBTB48 MIR4252 HES2 |
nsv545268 | Chr.1:6490316 - 6578004 on Build GRCh38 | Loss | TAS1R1 PLEKHG5 NOL9 |
nsv545269 | Chr.1:6546548 - 6630080 on Build GRCh38 | Loss | TAS1R1 NOL9 PHF13 THAP3 KLHL21 ZBTB48 |
nsv831536 | Chr.1:6429326 - 6605343 on Build GRCh38 | Loss | TAS1R1 PLEKHG5 ESPN NOL9 TNFRSF25 KLHL21 ZBTB48 MIR4252 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |