Assay Details
Target Gene Details
Entrez Gene ID: | 11240 |
Gene Name: | peptidyl arginine deiminase 2 |
Gene Aliases: |
PAD-H19, PAD2, PDI2 |
Location: |
Chr.1:17066761-17119453 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PADI2 | NM_007365.2 | NP_031391.2 | ||
XM_017000148.1 | XP_016855637.1 | |||
AB023211.1 | ||||
AB030176.1 | BAA82557.1 | |||
AK123164.1 | BAC85545.1 | |||
AK127006.1 | ||||
AK295783.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428421 | Chr.1:16437838 - 17157486 on Build GRCh38 | Gain+Loss | CROCCP2 LOC102723383 LOC105376805 LOC105376796 RNU1-3 SDHB RNU1-2 ATP13A2 MST1P2 NECAP2 NBPF1 LOC100132147 FAM231A CROCCP3 RNU1-1 SPATA21 RNU1-4 CROCC MIR3675 FAM231B PADI2 MFAP2 ESPNP MST1L |
nsv545690 | Chr.1:17040376 - 17159158 on Build GRCh38 | Gain | SDHB PADI2 |
nsv834280 | Chr.1:16888218 - 17123006 on Build GRCh38 | Loss | LOC105376805 SDHB CROCC RNU1-2 ATP13A2 PADI2 MFAP2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |