Assay Details
Target Gene Details
Entrez Gene ID: | 205 |
Gene Name: | adenylate kinase 4 |
Gene Aliases: |
AK 4, AK3, AK3L1, AK3L2 |
Location: |
Chr.1:65147342-65232145 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AK4 | NM_001005353.2 | NP_001005353.1 | ||
NM_013410.3 | NP_037542.1 | |||
NM_203464.2 | NP_982289.1 | |||
XM_017000612.1 | XP_016856101.1 | |||
XM_017000613.1 | XP_016856102.1 | |||
AK025926.1 | ||||
AK309057.1 | ||||
AK313611.1 | ||||
BC016180.1 | AAH16180.1 | |||
BC040224.1 | AAH40224.1 | |||
BC066944.1 | AAH66944.1 | |||
BC136886.1 | ||||
BC146653.1 | ||||
BC148270.1 | ||||
BG703687.1 | ||||
BM763432.1 | ||||
BU164554.1 | ||||
CR456830.1 | CAG33111.1 | |||
DB061266.1 | ||||
DR003868.1 | ||||
X60673.1 | CAA43088.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv173n100 | Chr.1:65102651 - 65215491 on Build GRCh38 | Gain | AK4 |
esv2676423 | Chr.1:64977641 - 65405798 on Build GRCh38 | Deletion | MIR101-1 DNAJC6 AK4 JAK1 LOC107983962 LINC01359 MIR3671 |
esv3586276 | Chr.1:65175235 - 65184831 on Build GRCh38 | Loss | AK4 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |