Assay Details
Target Gene Details
Entrez Gene ID: | 6146 |
Gene Name: | ribosomal protein L22 |
Gene Aliases: |
EAP, HBP15, HBP15/L22, L22 |
Location: |
Chr.1:6185020-6199619 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RPL22 | NM_000983.3 | NP_000974.1 | ||
AK097075.1 | ||||
AK311749.1 | ||||
BC035566.1 | AAH35566.1 | |||
BC058887.1 | AAH58887.1 | |||
BC066314.1 | AAH66314.1 | |||
CA336134.1 | ||||
CR456873.1 | CAG33154.1 | |||
D17652.1 | BAA04545.1 | |||
X59357.1 | CAA42007.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831425 | Chr.1:6129291 - 6264418 on Build GRCh38 | Loss | LINC00337 CHD5 LOC102724450 ICMT ACOT7 HES3 RPL22 RNF207 GPR153 |
nsv545236 | Chr.1:6161915 - 6262876 on Build GRCh38 | Loss | LINC00337 CHD5 LOC102724450 ICMT HES3 RPL22 RNF207 GPR153 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |