Assay Details
Target Gene Details
Entrez Gene ID: | 50651 |
Gene Name: | solute carrier family 45 member 1 |
Gene Aliases: |
DNB5 |
Location: |
Chr.1:8318085-8344167 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC45A1 | NM_001080397.2 | NP_001073866.2 | ||
XM_011541530.1 | XP_011539832.1 | |||
XM_011541531.1 | XP_011539833.1 | |||
AF118274.1 | AAD27583.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1012977 | Chr.1:8252522 - 8485210 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv516827 | Chr.1:8330828 - 8363450 on Build GRCh38 | Loss |
![]() ![]() |
nsv1743 | Chr.1:8317693 - 8351656 on Build GRCh38 | Insertion |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)