Assay Details
Target Gene Details
Entrez Gene ID: | 5915 |
Gene Name: | retinoic acid receptor beta |
Gene Aliases: |
HAP, MCOPS12, NR1B2, RARbeta1, RRB2 |
Location: |
Chr.3:24829323-25597932 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RARB | NM_001290216.2 | NP_001277145.1 | ||
DA240288.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3734 | Chr.3:24820077 - 24865375 on Build GRCh38 | Deletion |
![]() |
esv2759135 | Chr.3:24611022 - 24901435 on Build GRCh38 | Loss |
![]() |
nsv834643 | Chr.3:24738136 - 24901395 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)