Assay Details
Target Gene Details
Entrez Gene ID: | 80321 |
Gene Name: | centrosomal protein 70 |
Gene Aliases: |
BITE |
Location: |
Chr.3:138494339-138594383 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CEP70 | NM_001288964.1 | NP_001275893.1 | ||
NM_001288965.1 | NP_001275894.1 | |||
NM_001288967.1 | NP_001275896.1 | |||
NM_001320598.1 | NP_001307527.1 | |||
NM_024491.3 | NP_077817.2 | |||
XM_017007275.1 | XP_016862764.1 | |||
XM_017007282.1 | XP_016862771.1 | |||
XM_017007283.1 | XP_016862772.1 | |||
XM_017007284.1 | XP_016862773.1 | |||
AF202146.1 | AAG35616.1 | |||
AK023098.1 | BAB14403.1 | |||
AK294587.1 | ||||
AK299003.1 | ||||
AK301913.1 | ||||
AK314314.1 | ||||
BC030598.2 | AAH30598.1 | |||
BU634409.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv999041 | Chr.3:138429970 - 138629146 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv428422 | Chr.3:137344347 - 138686784 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3597916 | Chr.3:138478438 - 138638584 on Build GRCh38 | Gain |
![]() ![]() ![]() |
dgv4908n100 | Chr.3:138494324 - 139018846 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv522517 | Chr.3:138455681 - 138579140 on Build GRCh38 | Loss |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)