Assay Details
Target Gene Details
Entrez Gene ID: | 84172 |
Gene Name: | RNA polymerase I subunit B |
Gene Aliases: |
RPA135, RPA2, Rpo1-2 |
Location: |
Chr.2:112541915-112577153 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POLR1B | NM_001137604.2 | 4 | 1097 | NP_001131076.1 |
NM_001282772.1 | 6 | 915 | NP_001269701.1 | |
NM_001282774.1 | 5 | 1265 | NP_001269703.1 | |
NM_001282776.1 | 2 | 817 | NP_001269705.1 | |
NM_001282777.1 | 6 | 1357 | NP_001269706.1 | |
NM_001282779.1 | 5 | 1194 | NP_001269708.1 | |
NM_019014.5 | 5 | 1265 | NP_061887.2 | |
AK001678.1 | 5 | 737 | BAA91831.1 | |
AK022533.1 | 4 | 547 | BAB14082.1 | |
AK128044.1 | 5 | 1265 | BAC87247.1 | |
AK294027.1 | 5 | 666 | ||
AK301178.1 | 2 | 289 | ||
AK302814.1 | 6 | 915 | ||
BC110833.1 | 5 | 954 | AAI10834.1 | |
BX647360.1 | 6 | 1083 | ||
BX647683.1 | 5 | 644 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |