Assay Details
Target Gene Details
Entrez Gene ID: | 51280 |
Gene Name: | golgi membrane protein 1 |
Gene Aliases: |
C9orf155, GOLPH2, GP73, HEL46, PSEC0257, bA379P1.3 |
Location: |
Chr.9:86026141-86100201 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GOLM1 | NM_016548.3 | 10 | 2836 | NP_057632.2 |
NM_177937.2 | 10 | 2828 | NP_808800.1 | |
AF236056.1 | 10 | 2777 | AAF44663.1 | |
AI929234.1 | 1 | 255 | ||
AJ420432.1 | 1 | 1059 | ||
AK027717.1 | 10 | 2799 | ||
AK075542.1 | 10 | 2770 | BAC11685.1 | |
AK075557.1 | 10 | 2740 | ||
AK092943.1 | 11 | 2716 | ||
BC001740.1 | 10 | 2751 | AAH01740.1 | |
CA446701.1 | 1 | 265 | ||
EU794616.1 | 10 | 2795 |
Target Gene Details
Entrez Gene ID: | 60560 |
Gene Name: | N(alpha)-acetyltransferase 35, NatC auxiliary subunit |
Gene Aliases: |
EGAP, MAK10, MAK10P, bA379P1.1 |
Location: |
Chr.9:85941142-86026355 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAA35 | XM_011518903.2 | XP_011517205.1 | ||
XM_011518904.2 | XP_011517206.1 | |||
XM_017015012.1 | XP_016870501.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic non-DGV Variation |