Assay Details
Target Gene Details
Entrez Gene ID: | 116369 |
Gene Name: | solute carrier family 26 member 8 |
Gene Aliases: |
SPGF3, TAT1 |
Location: |
Chr.6:35943514-36024855 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC26A8 | NM_001193476.1 | 20 | 3156 | NP_001180405.1 |
NM_052961.3 | 20 | 3180 | NP_443193.1 | |
NM_138718.2 | 18 | 2903 | NP_619732.2 | |
XM_011514294.2 | 19 | 3321 | XP_011512596.1 | |
XM_017010235.1 | 20 | 3206 | XP_016865724.1 | |
AF314959.1 | 19 | 3050 | AAL26868.1 | |
AF331522.1 | 20 | 3124 | AAK95666.1 | |
AI015633.1 | 1 | 257 | ||
AK055314.1 | 10 | 2626 | ||
AK057276.1 | 10 | 2031 | BAB71408.1 | |
AK097410.1 | 14 | 2242 | ||
AK122928.1 | 20 | 3156 | ||
BC025408.2 | 18 | 2896 | AAH25408.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |