Assay Details
Target Gene Details
Entrez Gene ID: | 55862 |
Gene Name: | ethylmalonyl-CoA decarboxylase 1 |
Gene Aliases: |
HEL-S-76, MMCD, dJ351K20.2 |
Location: |
Chr.6:127288712-127343609 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ECHDC1 | NM_001002030.1 | 6 | 1895 | NP_001002030.1 |
NM_001105544.1 | 5 | 1673 | NP_001099014.1 | |
NM_001105545.1 | 4 | 1620 | NP_001099015.1 | |
NM_001139510.1 | 6 | 1673 | NP_001132982.1 | |
NM_018479.3 | 5 | 1842 | NP_060949.2 | |
XM_005267047.2 | 7 | 1820 | XP_005267104.1 | |
XM_005267048.1 | 6 | 1994 | XP_005267105.1 | |
XM_005267049.2 | 5 | 2109 | XP_005267106.1 | |
XM_005267050.2 | 6 | 1735 | XP_005267107.1 | |
XM_011535942.2 | 7 | 1791 | XP_011534244.1 | |
XM_017011056.1 | 7 | 1811 | XP_016866545.1 | |
AK025760.1 | 6 | 1645 | ||
AK025796.1 | 6 | 1673 | ||
AL834469.1 | 6 | 1834 | CAD39128.1 | |
BU684175.1 | 1 | 459 | ||
EU668355.1 | 4 | 1620 |
Target Gene Details
Entrez Gene ID: | 81847 |
Gene Name: | ring finger protein 146 |
Gene Aliases: |
- |
Location: |
Chr.6:127266340-127289893 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RNF146 | XM_006715571.3 | 5 | 3676 | XP_006715634.1 |
XM_011536161.2 | 6 | 3563 | XP_011534463.1 | |
XM_011536162.2 | 6 | 3615 | XP_011534464.1 | |
XM_011536163.2 | 6 | 3595 | XP_011534465.1 | |
XM_011536164.2 | 5 | 3454 | XP_011534466.1 | |
XM_017011336.1 | 3 | 3356 | XP_016866825.1 | |
XM_017011337.1 | 5 | 6597 | XP_016866826.1 | |
XM_017011338.1 | 7 | 3704 | XP_016866827.1 | |
XM_017011339.1 | 6 | 3733 | XP_016866828.1 | |
XM_017011340.1 | 6 | 3785 | XP_016866829.1 | |
XM_017011341.1 | 4 | 3484 | XP_016866830.1 | |
XM_017011342.1 | 5 | 3625 | XP_016866831.1 | |
XM_017011343.1 | 6 | 3734 | XP_016866832.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |