Assay Details
Target Gene Details
Entrez Gene ID: | 10099 |
Gene Name: | tetraspanin 3 |
Gene Aliases: |
TM4-A, TM4SF8, TSPAN-3 |
Location: |
Chr.15:77044017-77071228 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TSPAN3 | NM_001168412.1 | 6 | 1504 | NP_001161884.1 |
NM_005724.5 | 7 | 1696 | NP_005715.1 | |
NM_198902.2 | 6 | 1621 | NP_944492.1 | |
XM_017021857.1 | 7 | 1305 | XP_016877346.1 | |
AF133423.1 | 7 | 1496 | AAF08362.1 | |
AK001305.1 | 7 | 1542 | BAA91613.1 | |
AK001310.1 | 7 | 1528 | BAA91615.1 | |
AK001326.1 | 7 | 1639 | BAA91627.1 | |
AK027793.1 | 4 | 2747 | ||
AK075336.1 | 7 | 1577 | ||
AK075482.1 | 6 | 1502 | ||
AK298157.1 | 6 | 1385 | ||
BC000704.2 | 7 | 1597 | AAH00704.1 | |
BC004280.1 | 7 | 1558 | AAH04280.1 | |
BC009248.2 | 7 | 1561 | AAH09248.1 | |
BC011206.1 | 7 | 1590 | AAH11206.1 | |
M69023.1 | 3 | 970 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517068 | Chr.15:76962203 - 77088880 on Build GRCh38 | Gain+Loss |
![]() ![]() |
esv2760041 | Chr.15:76878209 - 77081106 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv833058 | Chr.15:76943554 - 77101298 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
esv2749898 | Chr.15:75764003 - 77933523 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1048833 | Chr.15:77002252 - 77046918 on Build GRCh38 | Gain |
![]() ![]() |
nsv1040852 | Chr.15:76867896 - 77845199 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1622 | Chr.15:77022478 - 77051924 on Build GRCh38 | Insertion |
![]() ![]() |
nsv509576 | Chr.15:76962019 - 77047336 on Build GRCh38 | Insertion |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)