Assay Details
Target Gene Details
Entrez Gene ID: | 3899 |
Gene Name: | AF4/FMR2 family member 3 |
Gene Aliases: |
LAF4, MLLT2-like |
Location: |
Chr.2:99545861-100142583 on Build GRCh38 |
Assay Gene Location: | Within Exon 33 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AFF3 | NM_001025108.1 | 24 | 7441 | NP_001020279.1 |
NM_002285.2 | 24 | 7458 | NP_002276.2 | |
XM_005263943.3 | 23 | 7335 | XP_005264000.2 | |
XM_005263945.3 | 15 | 6692 | XP_005264002.1 | |
XM_011511169.2 | 24 | 7438 | XP_011509471.1 | |
XM_011511170.2 | 25 | 7577 | XP_011509472.1 | |
XM_011511171.2 | 24 | 7393 | XP_011509473.1 | |
XM_011511172.2 | 23 | 7393 | XP_011509474.1 | |
XM_011511173.2 | 24 | 7454 | XP_011509475.1 | |
XM_011511174.2 | 24 | 7398 | XP_011509476.1 | |
XM_011511175.2 | 26 | 7688 | XP_011509477.1 | |
XM_011511176.2 | 23 | 7379 | XP_011509478.1 | |
XM_011511177.2 | 24 | 7469 | XP_011509479.1 | |
XM_011511178.2 | 14 | 6200 | XP_011509480.1 | |
XM_011511179.2 | 14 | 6197 | XP_011509481.1 | |
XM_017004085.1 | 15 | 6689 | XP_016859574.1 | |
XM_017004086.1 | 15 | 7170 | XP_016859575.1 | |
XM_017004087.1 | 15 | 7167 | XP_016859576.1 | |
AK055117.1 | 1 | 1301 | ||
AK091320.1 | 1 | 2419 | ||
BU740375.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |