Assay Details
Target Gene Details
Entrez Gene ID: | 27123 |
Gene Name: | dickkopf WNT signaling pathway inhibitor 2 |
Gene Aliases: |
DKK-2 |
Location: |
Chr.4:106921802-107036296 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DKK2 | NM_014421.2 | 4 | 1489 | NP_055236.1 |
AB033208.1 | 4 | 1006 | BAA85465.1 | |
AB033941.1 | 3 | 1346 | BAB20420.1 | |
AF177395.1 | 4 | 1501 | AAF02675.1 | |
AK309466.1 | 5 | 716 | ||
AK312695.1 | ||||
AY358414.1 | 4 | 994 | AAQ88780.1 | |
BC075077.2 | AAH75077.1 | |||
BC075078.2 | AAH75078.1 | |||
BC126328.1 | 4 | 1090 | ||
BC126330.1 | 4 | 1090 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv4453 | Chr.4:106904437 - 106934973 on Build GRCh38 | Insertion | DKK2 |
esv2752020 | Chr.4:106720427 - 106966306 on Build GRCh38 | Gain | DKK2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |