Assay Details
Target Gene Details
Entrez Gene ID: | 8653 |
Gene Name: | DEAD-box helicase 3, Y-linked |
Gene Aliases: |
DBY |
Location: |
Chr.Y:12903999-12920478 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDX3Y | NM_001122665.3 | NP_001116137.1 | ||
NM_001324195.1 | NP_001311124.1 | |||
NM_004660.4 | NP_004651.2 | |||
NR_136716.1 | ||||
NR_136717.1 | ||||
NR_136718.1 | ||||
NR_136719.1 | ||||
NR_136720.1 | ||||
NR_136721.1 | ||||
AF000984.1 | AAC51831.1 | |||
AF000985.1 | AAC51832.1 | |||
AK296366.1 | ||||
AK303176.1 | ||||
AK303638.1 | ||||
AK307884.1 | ||||
AK310382.1 | ||||
BC034942.2 | AAH34942.1 | |||
DC394594.1 | ||||
HY048505.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |