Assay Details
Target Gene Details
Entrez Gene ID: | 134728 |
Gene Name: | interleukin 1 receptor associated kinase 1 binding protein 1 |
Gene Aliases: |
AIP70, SIMPL |
Location: |
Chr.6:78867486-78979411 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IRAK1BP1 | NM_001010844.3 | 3 | 503 | NP_001010844.1 |
XM_006715336.3 | 3 | 544 | XP_006715399.1 | |
XM_011535447.1 | 3 | 543 | XP_011533749.1 | |
XM_011535448.2 | XP_011533750.1 | |||
XM_017010264.1 | 3 | 322 | XP_016865753.1 | |
XM_017010265.1 | XP_016865754.1 | |||
AK098678.1 | 3 | 1000 | ||
AK309003.1 | 3 | 440 | ||
BC112253.1 | 3 | 492 | AAI12254.1 | |
BC112255.1 | 3 | 492 | AAI12256.1 | |
BX103697.1 | 1 | 79 | ||
CR990080.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |