Assay Details
Target Gene Details
Entrez Gene ID: | 84869 |
Gene Name: | carbonyl reductase 4 |
Gene Aliases: |
SDR45C1 |
Location: |
Chr.4:168894484-169010317 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CBR4 | XM_005263315.2 | XP_005263372.1 | ||
XM_017008782.1 | XP_016864271.1 |
Target Gene Details
Entrez Gene ID: | 23022 |
Gene Name: | palladin, cytoskeletal associated protein |
Gene Aliases: |
CGI-151, CGI151, MYN, PNCA1, SIH002 |
Location: |
Chr.4:168497039-168928457 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PALLD | NM_001166108.1 | 22 | 4076 | NP_001159580.1 |
NM_001166109.1 | 19 | 3144 | NP_001159581.1 | |
NM_001166110.1 | 12 | 2643 | NP_001159582.1 | |
NM_016081.3 | 21 | 4025 | NP_057165.3 | |
XM_005262861.4 | 22 | 4729 | XP_005262918.1 | |
XM_005262866.2 | 21 | 4279 | XP_005262923.1 | |
XM_011531768.2 | 23 | 4983 | XP_011530070.1 | |
XM_011531769.2 | 22 | 4932 | XP_011530071.1 | |
XM_011531770.2 | 24 | 5029 | XP_011530072.1 | |
XM_011531771.2 | 21 | 4710 | XP_011530073.1 | |
XM_011531772.2 | 20 | 4611 | XP_011530074.1 | |
XM_011531773.1 | 22 | 4311 | XP_011530075.1 | |
XM_011531774.1 | 21 | 4260 | XP_011530076.1 | |
XM_011531775.1 | 21 | 3664 | XP_011530077.1 | |
XM_017007910.1 | 23 | 4978 | XP_016863399.1 | |
AB023209.1 | 12 | 2607 | ||
AF077041.1 | 8 | 2093 | AAD27774.1 | |
AF151909.1 | 8 | 1999 | AAD34146.1 | |
AF464873.1 | 21 | 4025 | AAL69964.1 | |
AL050093.1 | 3 | 728 | CAB43265.1 | |
BX537391.1 | 5 | 927 | CAD97633.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv34043 | Chr.4:168909873 - 169008517 on Build GRCh38 | Loss | CBR4 PALLD |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |