Assay Details
Target Gene Details
Entrez Gene ID: | 55471 |
Gene Name: | NADH:ubiquinone oxidoreductase complex assembly factor 7 |
Gene Aliases: |
C2orf56, MidA, PRO1853 |
Location: |
Chr.2:37231631-37259052 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFAF7 | NM_001083946.1 | 3 | 346 | NP_001077415.1 |
NM_144736.4 | 4 | 427 | NP_653337.1 | |
XM_005264410.1 | 4 | 407 | XP_005264467.1 | |
XM_005264411.1 | 4 | 405 | XP_005264468.1 | |
XM_011532956.1 | XP_011531258.1 | |||
AK294240.1 | 4 | 361 | ||
AK299015.1 | 4 | 401 | ||
AK302150.1 | 3 | 297 | ||
AK303555.1 | ||||
BC004548.2 | 4 | 392 | AAH04548.2 | |
BI334629.1 | 4 | 378 | ||
BM172474.1 | 4 | 432 | ||
BX538031.1 | 3 | 299 | CAD97976.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |