Assay Details
Target Gene Details
Entrez Gene ID: | 6677 |
Gene Name: | sperm adhesion molecule 1 |
Gene Aliases: |
HEL-S-96n, HYA1, HYAL1, HYAL3, HYAL5, PH-20, PH20, SPAG15 |
Location: |
Chr.7:123925232-123971410 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPAM1 | NM_001174044.1 | 3 | 742 | NP_001167515.1 |
NM_001174045.1 | 4 | 737 | NP_001167516.1 | |
NM_001174046.1 | 3 | 652 | NP_001167517.1 | |
NM_003117.4 | 3 | 563 | NP_003108.2 | |
NM_153189.2 | 3 | 563 | NP_694859.1 | |
AK292229.1 | 3 | 652 | ||
AY920278.1 | 3 | 738 | AAX13985.1 | |
AY920279.1 | 3 | 559 | AAX13986.1 | |
AY920280.1 | 4 | 712 | AAX13987.1 | |
AY920281.1 | 3 | 627 | AAX13988.1 | |
BC026163.1 | 3 | 486 | AAH26163.1 | |
BI831525.1 | 3 | 520 | ||
DB023094.1 | ||||
EU794686.1 | 3 | 526 | ||
L13781.1 | 1 | 225 | ||
S67798.1 | 3 | 496 | AAC60607.2 | |
X84347.1 | 2 | 430 | CAA59086.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |