Assay Details
Target Gene Details
Entrez Gene ID: | 4711 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit B5 |
Gene Aliases: |
CISGDH, SGDH |
Location: |
Chr.3:179604787-179624500 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFB5 | NM_001199957.1 | 4 | 570 | NP_001186886.1 |
NM_001199958.1 | 5 | 619 | NP_001186887.1 | |
NM_002492.3 | 6 | 726 | NP_002483.1 | |
AF047181.1 | 6 | 703 | AAC04266.1 | |
AW955118.1 | ||||
BC005271.1 | 6 | 717 | AAH05271.1 | |
BC009796.1 | 6 | 721 | AAH09796.1 | |
BC017909.1 | 6 | 716 | ||
BC093071.1 | 4 | 547 | AAH93071.1 | |
BX648034.1 | 4 | 564 | ||
CD364869.1 | 1 | 351 |
More Information
Additional Information:
For this assay, SNP(s) [rs149627280] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic non-DGV Variation |