Assay Details
Target Gene Details
Entrez Gene ID: | 493860 |
Gene Name: | coiled-coil domain containing 73 |
Gene Aliases: |
NY-SAR-79 |
Location: |
Chr.11:32602080-32794658 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCDC73 | NM_001008391.3 | 18 | 3453 | NP_001008392.2 |
XM_011520139.2 | 17 | 3314 | XP_011518441.1 | |
AK128159.1 | 9 | 2672 | BAC87304.1 | |
AK131470.1 | 18 | 3437 | BAD18615.1 |
Target Gene Details
Entrez Gene ID: | 10480 |
Gene Name: | eukaryotic translation initiation factor 3 subunit M |
Gene Aliases: |
B5, GA17, PCID1, TANGO7, hfl-B5 |
Location: |
Chr.11:32583767-32602873 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EIF3M | NM_001307929.1 | 8 | 1110 | NP_001294858.1 |
NM_006360.5 | 11 | 1506 | NP_006351.2 | |
AK292139.1 | ||||
AK304378.1 | 8 | 1046 | ||
BE935691.1 | 1 | 197 | ||
CK002866.1 | 2 | 448 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |