Assay Details
Target Gene Details
Entrez Gene ID: | 92799 |
Gene Name: | SH3KBP1 binding protein 1 |
Gene Aliases: |
PP203, Sb1 |
Location: |
Chr.19:40576851-40591399 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SHKBP1 | NM_138392.3 | 18 | 2137 | NP_612401.2 |
XM_006723474.1 | 18 | 2167 | XP_006723537.1 | |
XM_006723475.1 | 18 | 2164 | XP_006723538.1 | |
XM_011527492.1 | 15 | 1816 | XP_011525794.1 | |
XM_011527493.1 | 17 | 2063 | XP_011525795.1 | |
XM_017027473.1 | 18 | 2134 | XP_016882962.1 | |
XM_017027474.1 | 17 | 2033 | XP_016882963.1 | |
AF258553.1 | 18 | 2136 | AAG23756.1 | |
AK075057.1 | 18 | 2059 | BAC11374.1 | |
AK123695.1 | 16 | 2115 | ||
AK297006.1 | 12 | 1763 | ||
AK300808.1 | 14 | 1561 | ||
AK300819.1 | 17 | 1916 | ||
AK312748.1 | ||||
AY007117.1 | 9 | 1172 | ||
BC007653.1 | 10 | 1423 | AAH07653.1 | |
BC022855.1 | 18 | 2108 | AAH22855.1 | |
CA416700.1 | 1 | 243 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |