Assay Details
Target Gene Details
Entrez Gene ID: | 59340 |
Gene Name: | histamine receptor H4 |
Gene Aliases: |
AXOR35, BG26, GPCR105, GPRv53, H4, H4R, HH4R |
Location: |
Chr.18:24460568-24487427 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HRH4 | NM_001143828.1 | 1 | 77 | NP_001137300.1 |
NM_001160166.1 | 1 | 77 | NP_001153638.1 | |
NM_021624.3 | 1 | 77 | NP_067637.2 | |
XM_011526133.1 | 1 | 138 | XP_011524435.1 | |
AB044934.1 | BAB13698.1 | |||
AB045370.1 | 1 | 40 | BAB20091.1 | |
AF307973.1 | AAG32052.1 | |||
AF312230.1 | 1 | 77 | AAK12081.1 | |
AF325356.1 | AAL01684.1 | |||
AF329449.1 | AAK43542.1 | |||
AJ298292.1 | CAC83493.1 | |||
AY008280.1 | AAL09297.1 | |||
AY136745.1 | AAN01271.1 | |||
BC069136.1 | 1 | 77 | AAH69136.1 | |
BC112348.1 | 1 | 37 | AAI12349.1 | |
DQ835186.1 | ||||
DQ835187.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |