Assay Details
Target Gene Details
Entrez Gene ID: | 8894 |
Gene Name: | eukaryotic translation initiation factor 2 subunit beta |
Gene Aliases: |
EIF2, EIF2B, EIF2beta, PPP1R67, eIF-2-beta |
Location: |
Chr.20:34088298-34112356 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EIF2S2 | NM_001316363.1 | NP_001303292.1 | ||
NM_001316364.1 | NP_001303293.1 | |||
NM_003908.4 | NP_003899.2 | |||
XM_017028118.1 | XP_016883607.1 | |||
AB451237.1 | ||||
AB451362.1 | ||||
AK313025.1 | ||||
BC000461.2 | AAH00461.1 | |||
BC000934.2 | AAH00934.1 | |||
BC007888.2 | AAH07888.1 | |||
BT009795.1 | AAP88797.1 | |||
BX418731.2 | ||||
BX429051.2 | ||||
CR456734.1 | CAG33015.1 | |||
M29536.1 | AAA52383.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |