Assay Details
Target Gene Details
Entrez Gene ID: | 160 |
Gene Name: | adaptor related protein complex 2 alpha 1 subunit |
Gene Aliases: |
ADTAA, AP2-ALPHA, CLAPA1 |
Location: |
Chr.19:49766923-49807114 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AP2A1 | NM_014203.2 | NP_055018.2 | ||
NM_130787.2 | NP_570603.2 | |||
XM_011526556.2 | XP_011524858.1 | |||
XM_011526557.2 | XP_011524859.1 | |||
AK302005.1 | ||||
AL136925.1 | CAB66859.1 | |||
BC014214.2 | AAH14214.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | MIR4324 DHDH BAX RPS11 MIR6799 C19orf73 PIH1D1 C19orf68 CABP5 HRC LHB BSPH1 CGB8 ATF5 KCNJ14 SNAR-C1 BCAT2 SNORD34 CGB7 ZNF473 SNAR-A4 FLT3LG PPP1R15A LOC105372430 SNAR-A1 PLA2G4C-AS1 TMEM143 TEAD2 IZUMO1 PLA2G4C SPHK2 FAM83E MIR4751 SPACA4 FLJ26850 AKT1S1 LIG1 PRR12 ADM5 FGF21 PTH2 TRPM4 TSKS SNAR-A9 BCL2L12 SIGLEC16 SNAR-A12 RASIP1 LIN7B CCDC155 FUT1 GYS1 FTL SNAR-B1 CGB5 SNAR-A13 FUT2 SLC6A16 SCAF1 SNAR-C2 MIR4749 SNORD32A SNAR-B2 DBP MIR6798 SNAR-C3 CCDC114 MIR5088 NUP62 CA11 LOC101059948 SNAR-A10 CPT1C PTOV1 RUVBL2 HSD17B14 SNORD33 PRRG2 LOC101928295 SNAR-A5 TBC1D17 IL4I1 SULT2B1 LOC105447645 SNAR-C4 SIGLEC11 AP2A1 RCN3 DKKL1 MIR4750 IRF3 ELSPBP1 RPL18 TULP2 SNAR-A6 SNAR-A8 SNAR-A7 PNKP KCNA7 NTN5 VRK3 SNRNP70 MAMSTR GRIN2D NUCB1-AS1 NOSIP CARD8-AS1 LMTK3 PPFIA3 PRMT1 KDELR1 RRAS MIR6800 PTOV1-AS2 NUCB1 SNAR-A11 ALDH16A1 RPL13A MIR150 PLEKHA4 CGB3 MED25 FCGRT SNAR-G2 SNAR-D SYNGR4 SNAR-A3 CYTH2 SNAR-C5 FUZ CARD8 SNAR-G1 GRWD1 PTOV1-AS1 GFY SEC1P SLC17A7 SNORD35A CGB2 CD37 SNORD35B NTF4 LOC100287477 EMP3 ZNF114 CGB1 SNAR-A2 SNAR-A14 |
nsv470150 | Chr.19:49618821 - 49856789 on Build GRCh38 | Gain | PRR12 PTOV1-AS1 ADM5 CPT1C PTOV1 MED25 TSKS MIR4749 MIR6799 BCL2L12 PRMT1 AP2A1 RRAS MIR6800 PTOV1-AS2 IRF3 MIR5088 FUZ SCAF1 |
dgv53n68 | Chr.19:49724393 - 49943224 on Build GRCh38 | Loss | AKT1S1 PTOV1-AS1 PTOV1 MED25 TSKS MIR4749 MIR6799 TBC1D17 IL4I1 AP2A1 MIR6800 PTOV1-AS2 MIR4750 FUZ ATF5 MIR4751 NUP62 PNKP |
nsv953600 | Chr.19:49555044 - 49938043 on Build GRCh38 | Deletion | AKT1S1 PRR12 ADM5 CPT1C PTOV1 MED25 PRRG2 TSKS MIR6799 TBC1D17 IL4I1 BCL2L12 AP2A1 MIR4750 IRF3 FUZ ATF5 SCAF1 PNKP PTOV1-AS1 NOSIP MIR4749 PRMT1 RRAS MIR6800 PTOV1-AS2 MIR5088 MIR4751 NUP62 |
nsv1160655 | Chr.19:49489110 - 49841830 on Build GRCh38 | Deletion | MIR150 PRR12 ADM5 CPT1C MED25 SNORD33 PRRG2 FCGRT TSKS RPS11 MIR6799 BCL2L12 AP2A1 RCN3 IRF3 FUZ SCAF1 SNORD34 PTOV1-AS1 NOSIP SNORD35A SNORD32A PRMT1 RRAS MIR6800 SNORD35B MIR5088 RPL13A |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |