Assay Details
Target Gene Details
Entrez Gene ID: | 10054 |
Gene Name: | ubiquitin like modifier activating enzyme 2 |
Gene Aliases: |
ARX, HRIHFB2115, SAE2 |
Location: |
Chr.19:34428176-34469893 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UBA2 | NM_005499.2 | 10 | 1003 | NP_005490.1 |
XM_005258404.3 | 11 | 1284 | XP_005258461.2 | |
XM_006722962.2 | 9 | 894 | XP_006723025.1 | |
XM_011526304.2 | 11 | 1284 | XP_011524606.1 | |
XM_017026134.1 | 11 | 1284 | XP_016881623.1 | |
AF079566.1 | 10 | 1023 | AAD23914.1 | |
AF090384.1 | 10 | 969 | AAD12784.1 | |
AF110957.1 | 10 | 933 | AAD24434.1 | |
AK023120.1 | 10 | 1004 | ||
AK024114.1 | 2 | 178 | ||
AK124730.1 | 10 | 1144 | ||
AK299514.1 | 7 | 787 | ||
AK315521.1 | 10 | 980 | ||
AL136905.1 | 10 | 958 | CAB66839.1 | |
BC003153.1 | 10 | 981 | AAH03153.1 | |
BT009781.1 | 10 | 933 | AAP88783.1 | |
CR456756.1 | 10 | 933 | CAG33037.1 | |
U35832.1 | 10 | 958 | AAC99992.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv2469 | Chr.19:34442956 - 34476603 on Build GRCh38 | Insertion | UBA2 |
nsv1059395 | Chr.19:34430094 - 34499592 on Build GRCh38 | Gain | WTIP UBA2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |