Assay Details
Target Gene Details
Entrez Gene ID: | 10021 |
Gene Name: | hyperpolarization activated cyclic nucleotide gated potassium channel 4 |
Gene Aliases: |
SSS2 |
Location: |
Chr.15:73319859-73369264 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HCN4 | NM_005477.2 | NP_005468.1 | ||
XM_011521148.2 | XP_011519450.1 | |||
AJ132429.1 | CAB42604.1 | |||
AJ238850.1 | CAB52754.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv524049 | Chr.15:73320492 - 73387960 on Build GRCh38 | Loss | LOC107984728 HCN4 |
nsv569949 | Chr.15:73320036 - 73387960 on Build GRCh38 | Loss | LOC107984728 HCN4 |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion | HIGD2B LOC102723750 ISLR2 BBS4 C15orf59 FAM219B CLK3 LOXL1-AS1 C15orf59-AS1 NPTN CSK UBL7-AS1 UBL7 LOC105376731 LOC107984728 LOC283731 NPTN-IT1 ADPGK-AS1 NEO1 ADPGK MIR6881 CCDC33 C15orf39 TBC1D21 LOC732265 COX5A ULK3 STRA6 CYP1A2 ARID3B ISLR EDC3 PPCDC PML LOC101929333 GOLGA6C MIR4513 CPLX3 GOLGA6A SCAMP2 SEMA7A REC114 SCAMP5 CYP11A1 LOXL1 HCN4 STOML1 LOC729739 LMAN1L RPP25 CD276 CYP1A1 MPI MIR6882 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |