Assay Details
Target Gene Details
Entrez Gene ID: | 84904 |
Gene Name: | Rho guanine nucleotide exchange factor 39 |
Gene Aliases: |
C9orf100 |
Location: |
Chr.9:35658875-35665281 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARHGEF39 | NM_032818.2 | 9 | 2162 | NP_116207.2 |
XM_011518057.2 | 9 | 2163 | XP_011516359.1 | |
XM_017015225.1 | 8 | 2034 | XP_016870714.1 | |
XM_017015227.1 | 7 | 1809 | XP_016870716.1 | |
AK001187.1 | 4 | 1422 | ||
AK027548.1 | 7 | 1714 | BAB55192.1 | |
AY390225.1 | 9 | 2166 | AAR01010.1 | |
AY390226.1 | AAR01011.1 |
Target Gene Details
Entrez Gene ID: | 203260 |
Gene Name: | coiled-coil domain containing 107 |
Gene Aliases: |
PSEC0222 |
Location: |
Chr.9:35658282-35661503 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCDC107 | NM_001195200.1 | NP_001182129.1 | ||
NM_001195201.1 | NP_001182130.1 | |||
NM_001195217.1 | 5 | 672 | NP_001182146.1 | |
NM_174923.2 | 5 | 700 | NP_777583.2 | |
XM_005251403.4 | 5 | 683 | XP_005251460.1 | |
AK075523.1 | 5 | 658 | BAC11669.1 | |
BC018758.1 | 5 | 676 | AAH18758.1 | |
BC051285.1 | AAH51285.1 | |||
BE903559.1 | ||||
BM973388.1 | 1 | 573 | ||
DQ884403.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |