Assay Details
Target Gene Details
Entrez Gene ID: | 2263 |
Gene Name: | fibroblast growth factor receptor 2 |
Gene Aliases: |
BBDS, BEK, BFR-1, CD332, CEK3, CFD1, ECT1, JWS, K-SAM, KGFR, TK14, TK25 |
Location: |
Chr.10:121478330-121598656 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv428239 | Chr.10:120876200 - 121638984 on Build GRCh38 | Gain+Loss | LINC01153 FGFR2 WDR11 |
nsv470972 | Chr.10:121148384 - 121482279 on Build GRCh38 | Gain | LINC01153 FGFR2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |