Assay Details
Target Gene Details
Entrez Gene ID: | 29071 |
Gene Name: | C1GALT1 specific chaperone 1 |
Gene Aliases: |
C1GALT2, C38H2-L1, COSMC, HSPC067, MST143, TNPS |
Location: |
Chr.X:120625674-120630150 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C1GALT1C1 | NM_001011551.2 | 2 | 525 | NP_001011551.1 |
NM_152692.4 | 3 | 697 | NP_689905.1 | |
AB084170.1 | 2 | 390 | BAC41493.1 | |
AF161552.1 | 3 | 539 | AAF29039.1 | |
AF177284.1 | 2 | 400 | AAQ13670.1 | |
AJ238398.1 | 2 | 523 | CAC80277.1 | |
AK290111.1 | 2 | 406 | ||
AY159319.1 | 3 | 542 | AAN78129.1 | |
AY358642.1 | 2 | 366 | AAQ89005.1 | |
BC011930.2 | 2 | 344 | AAH11930.1 | |
BC050441.1 | 2 | 415 | AAH50441.1 | |
BG779539.1 | 2 | 500 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |