Assay Details
Target Gene Details
Entrez Gene ID: | 773 |
Gene Name: | calcium voltage-gated channel subunit alpha1 A |
Gene Aliases: |
APCA, BI, CACNL1A4, CAV2.1, EA2, FHM, HPCA, MHP, MHP1, SCA6 |
Location: |
Chr.19:13206442-13506460 on Build GRCh38 |
Assay Gene Location: | Within Exon 49 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CACNA1A | NM_000068.3 | 48 | 7716 | NP_000059.3 |
NM_001127221.1 | 47 | 7701 | NP_001120693.1 | |
NM_001127222.1 | 47 | 7703 | NP_001120694.1 | |
NM_001174080.1 | 48 | 7707 | NP_001167551.1 | |
NM_023035.2 | 48 | 7721 | NP_075461.2 | |
AB035727.2 | 47 | 7467 | BAA94766.2 | |
AF004883.1 | 48 | 7716 | AAB61612.1 | |
AF004884.1 | 48 | 7721 | AAB61613.1 | |
AF100774.1 | 19 | 2892 | AAC77460.1 | |
EU016197.1 | ||||
U79663.1 | 24 | 3531 | AAB49674.1 | |
U79664.1 | 24 | 3531 | AAB49675.1 | |
U79665.1 | 24 | 3536 | AAB49676.1 | |
U79666.1 | 47 | 7687 | AAB64179.1 | |
U79667.1 | 23 | 3500 | AAB49677.1 | |
U79668.1 | AAB49678.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |