Assay Details
Target Gene Details
Entrez Gene ID: | 55008 |
Gene Name: | HECT and RLD domain containing E3 ubiquitin protein ligase family member 6 |
Gene Aliases: |
- |
Location: |
Chr.4:88378565-88443097 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HERC6 | NM_001165136.1 | 2 | 426 | NP_001158608.1 |
NM_017912.3 | 2 | 426 | NP_060382.3 | |
XM_005263083.3 | 2 | 623 | XP_005263140.1 | |
XM_011532053.2 | 2 | 625 | XP_011530355.1 | |
XM_017008333.1 | 2 | 106 | XP_016863822.1 | |
AF336798.1 | 2 | 426 | AAQ14893.1 | |
AK092403.1 | 2 | 174 | BAC03879.1 | |
AK295832.1 | 2 | 336 | ||
AY653201.1 | 2 | 426 | AAV66895.1 | |
AY653202.1 | 2 | 426 | AAV66896.1 | |
AY653203.1 | 2 | 426 | AAV66897.1 | |
BC042047.1 | 2 | 302 | AAH42047.2 | |
BX647121.1 | 2 | 102 | CAI46056.1 | |
DA221524.1 | 3 | 217 | ||
DC344051.1 | 3 | 387 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |