Assay Details
Target Gene Details
Entrez Gene ID: | 8659 |
Gene Name: | aldehyde dehydrogenase 4 family member A1 |
Gene Aliases: |
ALDH4, P5CD, P5CDh |
Location: |
Chr.1:18871430-18902799 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ALDH4A1 | NM_001161504.1 | 5 | 533 | NP_001154976.1 |
NM_001319218.1 | 5 | 673 | NP_001306147.1 | |
NM_003748.3 | 5 | 673 | NP_003739.2 | |
NM_170726.2 | 5 | 673 | NP_733844.1 | |
AK222486.1 | 5 | 429 | BAD96206.1 | |
AK289972.1 | 5 | 599 | ||
AK294552.1 | 5 | 533 | ||
BC007581.1 | 5 | 403 | AAH07581.1 | |
BC023600.2 | 5 | 414 | AAH23600.1 | |
BM856369.1 | 5 | 452 | ||
FJ462711.1 | 5 | 411 | ||
U24266.1 | 5 | 427 | AAC50500.1 | |
U24267.1 | 5 | 427 | AAC50501.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs113846237] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Exonic non-DGV Variation |