Assay Details
Target Gene Details
Entrez Gene ID: | 3065 |
Gene Name: | histone deacetylase 1 |
Gene Aliases: |
GON-10, HD1, RPD3, RPD3L1 |
Location: |
Chr.1:32292103-32333628 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HDAC1 | NM_004964.2 | NP_004955.2 | ||
XM_011541309.2 | XP_011539611.1 | |||
AB451297.1 | ||||
AB451430.1 | ||||
AK225555.1 | ||||
AK296308.1 | ||||
AK299792.1 | ||||
AK313061.1 | ||||
AY627042.1 | AAT44863.1 | |||
BC000301.2 | AAH00301.1 | |||
CR541717.1 | CAG46518.1 | |||
D50405.1 | BAA08909.1 | |||
U50079.1 | AAC50475.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |