Assay Details
Target Gene Details
Entrez Gene ID: | 79368 |
Gene Name: | Fc receptor like 2 |
Gene Aliases: |
CD307b, FCRH2, IFGP4, IRTA4, SPAP1, SPAP1A, SPAP1B, SPAP1C |
Location: |
Chr.1:157745733-157777472 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FCRL2 | NM_001159488.1 | 9 | 1683 | NP_001152960.1 |
NM_030764.3 | 12 | 1864 | NP_110391.2 | |
NR_125358.1 | 8 | 946 | ||
XM_011509974.2 | 12 | 2225 | XP_011508276.1 | |
XM_011509975.2 | 11 | 2161 | XP_011508277.1 | |
XM_011509976.2 | 11 | 2134 | XP_011508278.1 | |
XM_017002316.1 | 11 | 2170 | XP_016857805.1 | |
XM_017002317.1 | 11 | 1973 | XP_016857806.1 | |
XM_017002318.1 | 10 | 1893 | XP_016857807.1 | |
XM_017002319.1 | 10 | 1885 | XP_016857808.1 | |
AF319438.1 | AAK01402.1 | |||
AF459633.1 | 12 | 1852 | AAL60249.1 | |
AL833194.1 | 7 | 3723 | ||
AL833361.1 | 2 | 2384 | ||
AY043465.1 | 12 | 1851 | AAK91778.1 | |
AY358130.1 | AAQ88497.1 | |||
BC069185.1 | 8 | 950 | AAH69185.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |