Assay Details
Target Gene Details
Entrez Gene ID: | 8829 |
Gene Name: | neuropilin 1 |
Gene Aliases: |
BDCA4, CD304, NP1, NRP, VEGF165R |
Location: |
Chr.10:33177491-33334905 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NRP1 | NM_001244972.1 | 17 | 4488 | NP_001231901.1 |
NM_001244973.1 | 17 | 4485 | NP_001231902.1 | |
NM_003873.5 | 17 | 4506 | NP_003864.4 | |
NR_045259.1 | 16 | 4303 | ||
XM_006717521.1 | 17 | 4283 | XP_006717584.1 | |
XM_006717522.1 | 16 | 4232 | XP_006717585.1 | |
XM_006717523.2 | 16 | 4229 | XP_006717586.1 | |
XM_006717524.1 | 16 | 4178 | XP_006717587.1 | |
XM_006717525.1 | 16 | 4175 | XP_006717588.1 | |
XM_017016865.1 | 16 | 4208 | XP_016872354.1 | |
XM_017016866.1 | 16 | 4100 | XP_016872355.1 | |
AB209641.1 | 17 | 4134 | BAD92878.1 | |
AL833608.1 | 16 | 4034 | CAH10397.1 | |
BX510902.1 | 17 | 4170 | CAD91133.1 | |
BX648250.1 | 17 | 4454 | ||
CR749333.1 | 17 | 4507 | CAH18187.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831829 | Chr.10:33178304 - 33344113 on Build GRCh38 | Loss | LOC105376488 NRP1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |